CAS RN 1953-02-2
IUPAC Namn:
2-(2-sulfanylpropanamido)acetic acid
Synonymer:
α mercaptopropionylglycine
Molekylvikt (g/mol):
163.19
Molekylär sammansättning:
C5H9NO3S
InChi Key:
YTGJWQPHMWSCST-UHFFFAOYNA-N
SMILES:
CC(S)C(=O)NCC(O)=O
1
–
3
av
3
Resultat
1
Tiopronin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tiopronin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tiopronin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.