CAS RN 29882-07-3
IUPAC Namn:
4-chloro-1,1-dimethoxybutane
Synonymer:
4-chloro-1,1-dimethoxybutane
Molekylvikt (g/mol):
152.62
Molekylär sammansättning:
C6H13ClO2
InChi Key:
LTLKJYMNUSSFAH-UHFFFAOYSA-N
SMILES:
COC(CCCCl)OC
1
–
4
av
4
Resultat
1
4-Chloro-1,1-dimethoxybutane (4-Chlorobutyraldehyde Dimethyl Acetal), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-Chlorobutanal Dimethyl Acetal 90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Chlorobutanal Dimethyl Acetal 90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
4-Chlorobutanal Dimethyl Acetal 90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.