CAS RN 400038-68-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
2-Keto Zolpidem, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Oxozolpidem (N,N,6-Trimethyl-2-(4-methylphenyl)-alpha-oxo-imidazo[1,2-a]pyridine-3-acetamide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
2-Keto Zolpidem, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-Keto Zolpidem, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.