CAS RN 42540-40-9
IUPAC Namn:
sodium (6R,7R)-7-[(2R)-2-(formyloxy)-2-phenylacetamido]-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonymer:
cefamandole nafate cephamandole nafate O-formylcefamandole sodium cefamandol nafato
Molekylvikt (g/mol):
512.49
Molekylär sammansättning:
C19H17N6NaO6S2
InChi Key:
ICZOIXFFVKYXOM-YCLOEFEOSA-M
SMILES:
[Na+].CN1N=NN=C1SCC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](OC=O)C1=CC=CC=C1)C2=O)C([O-])=O
1
–
3
av
3
Resultat
1
Cefamandole Nafate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cefamandole Nafate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Cefamandole nafate, MedChemExpress
MedChemExpress Cefamandole nafate (Cefamandole formate sodium) is a second-generation broad-spectrum cephalosporin antibiotic.