CAS RN 52378-40-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
N-Cyano-N’-[2-[(4-methyl-5-imidazolyl)methylthio]ethyl]-S-methylisothiourea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cimetidine EP Impurity A, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
N-Cyano-N’-[2-[(4-methyl-5-imidazolyl)methylthio]ethyl]-S-methylisothiourea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
N-Cyano-N’-[2-[(4-methyl-5-imidazolyl)methylthio]ethyl]-S-methylisothiourea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.