CAS RN 528-53-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
5
av
5
Resultat
1
Delphinidin Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Delphinidin Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Delphinidin Chloride (~80% purity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Delphinidin Chloride (~80% purity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5