CAS RN 55974-42-0
IUPAC Namn:
2-{2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl}-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione dihydrochloride
Synonymer:
2-{2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl}-4,4-dimethylisoquinoline-1,3-dione dihydrochloride
Molekylvikt (g/mol):
480.43
Molekylär sammansättning:
C24H31Cl2N3O3
InChi Key:
SAJKHRHHDGSJEZ-UHFFFAOYSA-N
SMILES:
Cl.Cl.COC1=CC=CC=C1N1CCN(CCN2C(=O)C3=CC=CC=C3C(C)(C)C2=O)CC1
1
–
3
av
3
Resultat
1
ARC 239 Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ ARC 239 dihydrochloride
α2B antagonist
3
ARC 239 Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.