CAS RN 6802-95-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
2,2-Diphenylhydantoic Acid, TRC
CAS: 6802-95-5 Molekylformel: C15 H14 N2 O3 Molekylvikt (g/mol): 270.28 Synonym: (Carbamoylamino)-(diphenyl)acetic Acid,Phenytoin Imp. E (EP),Phenytoin Sodium Imp. E (EP) IUPAC-namn: 2-(carbamoylamino)-2,2-diphenylacetic acid LEDER: NC(=O)NC(C(=O)O)(c1ccccc1)c2ccccc2
2
(Carbamoylamino)(diphenyl)acetic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.