CAS RN 717906-29-1
IUPAC Namn:
N-methyl-N-{2-[({2-[(2-oxo-2,3-dihydro-1H-indol-5-yl)amino]-5-(trifluoromethyl)pyrimidin-4-yl}amino)methyl]phenyl}methanesulfonamide
Synonymer:
N-methyl-N-{2-[({2-[(2-oxo-1,3-dihydroindol-5-yl)amino]-5-(trifluoromethyl)pyrimidin-4-yl}amino)methyl]phenyl}methanesulfonamide
Molekylvikt (g/mol):
506.5
Molekylär sammansättning:
C22H21F3N6O3S
InChi Key:
POJZIZBONPAWIV-UHFFFAOYSA-N
SMILES:
CN(C1=CC=CC=C1CNC1=NC(NC2=CC3=C(NC(=O)C3)C=C2)=NC=C1C(F)(F)F)S(C)(=O)=O
1
–
4
av
4
Resultat
1
PF 431396, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
PF 431396, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
PF 431396, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.