CAS RN 82648-70-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(E)-(3-Cyano-2-methyl-2-propenyl)-phosphonic Acid Diethyl Ester (~10% Z-isomer), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(E)-(3-Cyano-2-methyl-2-propenyl)-phosphonic Acid Diethyl Ester (~10% Z-isomer), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(E)-(3-Cyano-2-methyl-2-propenyl)-phosphonic Acid Diethyl Ester (~10% Z-isomer), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.