missing translation for 'onlineSavingsMsg'
Learn More
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97% Product Code.: 15204288
compliance-icons

1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%

Product Code. 15204288 Shop All Thermo Scientific Chemicals Products
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15204288

Brand: Thermo Scientific Acros 452690050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84322-56-5 | C6F3I3 | 509.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84322-56-5
Molecular Formula C6F3I3
Molecular Weight (g/mol) 509.78
InChI Key NTAZOPPTLZSXQH-UHFFFAOYSA-N
Synonym 1,3,5-trifluoro-2,4,6-triiodo-benzene, 1,3,5-trilfluoro-2,4,6-triiodobenzene, benzene,1,3,5-trifluoro-2,4,6-triiodo, pubchem10637, 1,3,5-trifluoro-2,4, 1,3,5-triiodotrifluorobenzene, 1,3,5-trifluoro-2,4,6-triiodob, benzene, 1,3,5-trifluoro-2,4,6-triiodoShow MoreShow Less
PubChem CID 3852430
IUPAC Name 1,3,5-trifluoro-2,4,6-triiodobenzene
SMILES C1(=C(C(=C(C(=C1I)F)I)F)I)F

Specifications

Boiling Point 154.0°C to 156.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 509.78
Percent Purity 97%
Chemical Name or Material 1,3,5-Trifluoro-2,4,6-triiodobenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number345321 Certificate TypeCertificate of Analysis Date02/04/2024 Product Code452690050,452690010
Lot Number345065 Certificate TypeCertificate of Analysis Date08/02/2023 Product Code452690050,452690010
Lot Number344182 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code452690050,452690010
Lot Number343891 Certificate TypeCertificate of Analysis Date17/10/2022 Product Code452690050,452690010
Lot Number341114 Certificate TypeCertificate of Analysis Date15/02/2022 Product Code452690050,452690010
compliance-icons
Product Identifier
  • 1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.