Learn More
1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one, 97%, Thermo Scientific Chemicals Product Code.: 11976291
1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one, 97%, Thermo Scientific Chemicals
Product Code. 11976291 Shop All Thermo Scientific Chemicals Products
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11976291
Brand: Thermo Scientific Acros 135910050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2147-83-3
Molecular Formula C12H14N3O
Molecular Weight (g/mol) 216.26
MDL Number MFCD00044827
InChI Key YFEOSTXFQCDCAR-UHFFFAOYSA-O
Synonym 1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazole-2-one, 1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazol-2-one, 3-1,2,3,6-tetrahydropyridin-4-yl-1h-benzimidazol-2-one, 1-1,2,3,6-tetrahydro-4-pyridyl-2-benzimidazolinone, 1-1,2,3,6-tetrahydropyridin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-4-1,2,5,6-tetrahydropyridyl-3-hydrobenzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-3h-1,3-benzodiazol-2-one, benzoimidazol-2-one, 1-1,2,3,6-tetrahydro-pyridin-4-yl-1,3-dihydro
PubChem CID 75070
IUPAC Name 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-benzimidazol-2-one
SMILES O=C1NC2=CC=CC=C2N1C1=CC[NH2+]CC1
Specifications
Melting Point 195.0°C to 200.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,dichloromethane and acetone
Formula Weight 215.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0002340 Certificate TypeCertificate of Analysis Date28/06/2021 Product Code135910010,135910050
Lot NumberA0120631 Certificate TypeCertificate of Analysis Date15/09/2020 Product Code
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Product Title
1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one, 97%, Thermo Scientific Chemicals >