missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroethyl chloroformate, 98%, Thermo Scientific Chemicals

CAS: 50893-53-3 | C3H4Cl2O2 | 142.96 g/mol

391.00 SEK - 1370.00 SEK

Chemical Identifiers

CAS 50893-53-3
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.96
MDL Number MFCD00000647
InChI Key QOPVNWQGBQYBBP-UHFFFAOYNA-N
Synonym 1-chloroethyl chloroformate, chloroformic acid 1-chloroethyl ester, 1-chloro ethyl chloroformate, 1-chloroethylchloroformate, carbonochloridic acid, 1-chloroethyl ester, ace-cl, alpha-chloroethyl chloroformate, chloro 1-chloroethoxy methanone, .alpha.-chloroethyl chloroformate, pubchem17418
PubChem CID 521305
IUPAC Name 1-chloroethyl carbonochloridate
SMILES CC(Cl)OC(Cl)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339045
SDS View Documents
Thermo Scientific Alfa Aesar
L14197.06
5 g
391.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11349045
SDS View Documents
Thermo Scientific Alfa Aesar
L14197.14
25 g
1370.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Chloroethyl chloroformate acts as a reagent in the selective N-dealkylation of tertiary amines. It is also used as a reactant in the preparation of alkyl 1-chloroethyl carbonates. Further, it is used for N-demethylation reaction in the determination of multiple drugs of abuse in biological fluids using capillary electrophoresis with native fluorescence and laser-induced fluorescence detection. It plays an important role as a catalyst for the chemoselective desilylation of silyl-protected alcohols. In addition to this, it is used to cleave benzhydryl groups from amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloroethyl chloroformate acts as a reagent in the selective N-dealkylation of tertiary amines. It is also used as a reactant in the preparation of alkyl 1-chloroethyl carbonates. Further, it is used for N-demethylation reaction in the determination of multiple drugs of abuse in biological fluids using capillary electrophoresis with native fluorescence and laser-induced fluorescence detection. It plays an important role as a catalyst for the chemoselective desilylation of silyl-protected alcohols. In addition to this, it is used to cleave benzhydryl groups from amines.

Solubility
Miscible with most organic solvents.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agent, strong base, alcohols and amines.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H314-H318-H226-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302-H314-H330

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, FLAMMABLE, N.O.S.

missing translation for 'einecsNumber' : 256-834-2

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chloroethyl chloroformate, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.