Learn More
2-Methoxytetrahydropyran, 98%
Product Code. 15229656 Shop All Thermo Scientific Chemicals Products
25 g, 25g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
CAS: 6581-66-4 | C6H12O2 | 116.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6581-66-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key XTDKZSUYCXHXJM-UHFFFAOYSA-N
Synonym 2-methoxytetrahydropyran, tetrahydro-2-methoxy-2h-pyran, pyran, 2-methoxy-tetrahydro, 2h-pyran,tetrahydro-2-methoxy, 2h-pyran, tetrahydro-2-methoxy, 2-methoxytetrahydro-2h-pyran, methoxytetrahydropyran, 2-methoxyoxan-3-yl, 5-17-03-00078 beilstein handbook reference
PubChem CID 23057
IUPAC Name 2-methoxyoxane
SMILES COC1CCCCO1
Specifications
Density 0.9630g/mL
Boiling Point 128°C to 129°C
Flash Point 25°C
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 0.963
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 2-Methoxytetrahydropyran
compliance-icons
Product Identifier
  • 2-Methoxytetrahydropyran, 98%
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Product Content Correction
Product Title
2-Methoxytetrahydropyran, 98% >