missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dibromobenzoic acid, 99%, Thermo Scientific™ Product Code.: 15565076

3,5-Dibromobenzoic acid, 99%, Thermo Scientific™

Product Code. 15565076 Shop All Thermo Scientific Chemicals Products
25 g, 25g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15565076

Brand: Thermo Scientific Acros 352490250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 618-58-6
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00051758
InChI Key SFTFNJZWZHASAQ-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3hShow MoreShow Less
PubChem CID 12063
IUPAC Name 3,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 212.0°C to 214.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Linear Formula Br2C6H3CO2H
Quantity 25 g
Formula Weight 279.91
Percent Purity 99%
Chemical Name or Material 3,5-Dibromobenzoic acid
compliance-icons
Product Identifier
  • 3,5-Dibromobenzoic acid, 99%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dibromobenzoic acid, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.