Learn More
Lenz Laborglasintrumente 3-Way Stopcocks, with PTFE Plug
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
628.00 SEK - 807.00 SEK
Specifications
Material Borosilicate Glass (3.3)
Includes PTFE plug
View More Specs
Products 2
Product Code Brand Plug Bore Price Quantity & Availability  
Product Code Brand Plug Bore Price Quantity & Availability  
10519971
View Documents
Lenz Laborglasintrumente
02541804
4 mm
807.00 SEK
Each
Only null left
Estimated Shipment: 02-09-2025
Add to basket
10523641
View Documents
Lenz Laborglasintrumente
02541402
2.5 mm
628.00 SEK
Each
Only null left
Estimated Shipment: 19-08-2025
Add to basket
Description
Description
  • Acc. to DIN 12554 and in addition to DIN standard
  • T-bore
  • With PTFE plug and screwthread retaining device
  • Material: borosilicate glass 3.3
  • Leakage rate to DIN 12540-1
Specifications
Specifications
Borosilicate Glass (3.3)
PTFE plug
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasintrumente 3-Way Stopcocks, with PTFE Plug