missing translation for 'onlineSavingsMsg'
Learn More
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran, Thermo Scientific™ Product Code.: 10109419

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran, Thermo Scientific™

Product Code. 10109419
1 g, 1g
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10109419

Brand: Thermo Scientific Maybridge CC29939DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1131912-76-9
Molecular Formula C11H21BO3
Molecular Weight (g/mol) 212.10
MDL Number MFCD11506065
InChI Key NLSMOSUUBUCSPL-UHFFFAOYSA-N
Synonym tetrahydropyran-4-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-2h-pyran-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl tetrahydropyran, tetrahydropyran-4-boronic acid, pinacol ester, tetrahydropyran-4-boronicacidpinacolester, 4,4,5,5-tetramethyl-2-oxan-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl oxane, tetrahydro-2h-pyran-4-yl boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydro-2h-pyran
PubChem CID 42614649
IUPAC Name 4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CCOCC1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 212.09
Percent Purity 97%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.