missing translation for 'onlineSavingsMsg'
Learn More
4',4″-Dibromodiphenylbutadiyne, 97%, Thermo Scientific™ Product Code.: 15214418

4',4″-Dibromodiphenylbutadiyne, 97%, Thermo Scientific™

Product Code. p-7077520 Shop All Thermo Scientific Chemicals Products
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15214418

Brand: Thermo Scientific Acros 454912500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 959-88-6
Molecular Formula C16H8Br2
Molecular Weight (g/mol) 360.05
MDL Number MFCD00825678
InChI Key IIERBCCHBFZMQZ-UHFFFAOYSA-N
Synonym 1,4-bis 4-bromophenyl-1,3-butadiyne, 1,3-butadiyne, 1,4-di 4-bromophenyl, 4,4'-dibromodiphenylbutadiyne, 1,4-bis 4-bromophenyl buta-1,3-diyne, 1,1'-buta-1,3-diyne-1,4-diyl bis 4-bromobenzene, 1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene, 1-bromo-4-4-4-bromophenyl buta-1,3-diynyl benzene, 1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene #, 1-bromo-4-4-4-bromophenyl buta-1,3-diyn-1-yl benzene
PubChem CID 632304
IUPAC Name 1-bromo-4-[4-(4-bromophenyl)buta-1,3-diynyl]benzene
SMILES BrC1=CC=C(C=C1)C#CC#CC1=CC=C(Br)C=C1

Specifications

Melting Point 264.0°C to 265.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 250 mg
Formula Weight 360.05
Percent Purity 97%
Chemical Name or Material 4',4''-Dibromodiphenylbutadiyne
compliance-icons
Product Identifier
  • 4',4''-Dibromodiphenylbutadiyne, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4',4″-Dibromodiphenylbutadiyne, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.