Learn More
4-[6-(Tributylstannyl)-2-pyridinyl]morpholine, 95%, Thermo Scientific™
1690.00 SEK - 4365.00 SEK
Chemical Identifiers
CAS | 869901-24-6 |
---|---|
Molecular Formula | C21H38N2OSn |
Molecular Weight (g/mol) | 453.258 |
MDL Number | MFCD08271894 |
InChI Key | LDERBIPTTICDSG-UHFFFAOYSA-N |
Synonym | 4-6-tributylstannyl pyridin-2-yl morpholine, 4-6-tributylstannyl-2-pyridinyl morpholine, pubchem21425, 4-6-tributylstannanyl-pyridin-2-yl-morpholine, morpholine,4-6-tributylstannyl-2-pyridinylShow More |
PubChem CID | 18525739 |
IUPAC Name | tributyl-(6-morpholin-4-ylpyridin-2-yl)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)N2CCOCC2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10012264
|
Thermo Scientific Maybridge
CC38341CB |
250 mg | Amber Glass Bottle |
1690.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10715213
|
Thermo Scientific Maybridge
CC38341DA |
1 g | Amber Glass Bottle |
4365.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
869901-24-6 | |
453.258 | |
LDERBIPTTICDSG-UHFFFAOYSA-N | |
18525739 | |
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)N2CCOCC2 |
C21H38N2OSn | |
MFCD08271894 | |
4-6-tributylstannyl pyridin-2-yl morpholine, 4-6-tributylstannyl-2-pyridinyl morpholine, pubchem21425, 4-6-tributylstannanyl-pyridin-2-yl-morpholine, morpholine,4-6-tributylstannyl-2-pyridinylShow More | |
tributyl-(6-morpholin-4-ylpyridin-2-yl)stannane |
Specifications
869901-24-6 | |
100.0 | |
C21H38N2OSn | |
4-6-tributylstannyl pyridin-2-yl morpholine, 4-6-tributylstannyl-2-pyridinyl morpholine, pubchem21425, 4-6-tributylstannanyl-pyridin-2-yl-morpholine, morpholine,4-6-tributylstannyl-2-pyridinyl | |
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)N2CCOCC2 | |
453.258 | |
453.25 | |
4-[6-(Tributylstannyl)-2-pyridinyl]morpholine |
94.5 | |
95% | |
MFCD08271894 | |
LDERBIPTTICDSG-UHFFFAOYSA-N | |
tributyl-(6-morpholin-4-ylpyridin-2-yl)stannane | |
18525739 | |
95% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger