Learn More
Gilac™ 40 L Ingredients Container with Lid
40 L Ingredients Container with Lid guarantees hygienic storage of ingredients.
459.00 SEK - 703.00 SEK
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
17908841
|
Gilac™
G144621 |
White |
459.00 SEK
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||||
17998931
|
Gilac™
G144825 |
Clear |
703.00 SEK
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||||
Description
- Guarantees hygienic storage of ingredients (with lid).
- Easily accessible.
- Completely smooth bottom with no visible screws.
- Rounded corners and completely smooth walls for easy cleaning.
- Container stackable (with optional lid) and nestable.
- Compatible with GILAC universal 600 x 400 rolling dollies for easy handling.
- The 40 L ingredients container can be stacked on a 80 L ingredients container, on a rectangular or a 600 x 400 HACCP container.
- Wide range of colored lids are available.
- A single range of lids for 40 L and 80 L ingredients containers.
- Container designed in slightly transparent HDPE, allowing the content and level of material to be seen.
Specifications
Storage Tote | |
HDPE | |
Rectangular |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Gilac™ 40 L Ingredients Container with Lid