missing translation for 'onlineSavingsMsg'
Learn More
5-Hydroxy-2-nitrobenzaldehyde, 98% Product Code.: 15575036

5-Hydroxy-2-nitrobenzaldehyde, 98%

Product Code. 15575036 Shop All Thermo Scientific Chemicals Products
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 15575036

Brand: Thermo Scientific Acros 121650050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 42454-06-8 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42454-06-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007332
InChI Key XLYPHUGUKGMURE-UHFFFAOYSA-N
Synonym 3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
PubChem CID 39211
IUPAC Name 5-hydroxy-2-nitrobenzaldehyde
SMILES OC1=CC=C(C(C=O)=C1)[N+]([O-])=O

Specifications

Melting Point 168.0°C to 170.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula HOC6H3(NO2)CHO
Quantity 5 g
Beilstein 08,63
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 5-Hydroxy-2-nitrobenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number343968 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code121650250,121650010,121650050
Lot Number344071 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code121650250,121650010,121650050
Lot Number344310 Certificate TypeCertificate of Analysis Date18/11/2021 Product Code121650250,121650010,121650050
compliance-icons
Product Identifier
  • 5-Hydroxy-2-nitrobenzaldehyde, 98%
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Hydroxy-2-nitrobenzaldehyde, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.