Learn More
5-Methoxytryptamine hydrochloride, 97%, Thermo Scientific™ Product Code.: 10386670
5-Methoxytryptamine hydrochloride, 97%, Thermo Scientific™
Product Code. p-158717 Shop All Thermo Scientific Chemicals Products
250 mg, Glass bottle, 250mg
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
Product Code. 10386670
Brand: Thermo Scientific Acros 126132500
Request Bulk or Custom Format
Chemical Identifiers
CAS 66-83-1
Molecular Formula C11H14N2O·HCl
Molecular Weight (g/mol) 226.71
MDL Number MFCD00012684
InChI Key TXVAYRSEKRMEIF-UHFFFAOYSA-N
Synonym 5-methoxytryptamine hydrochloride, mexamine hydrochloride, o-methylserotonin hydrochloride, 2-5-methoxy-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methoxyindole hydrochloride, unii-4265ib6qek, 5-methoxy-1h-indole-3-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-methoxy-, monohydrochloride, 5-methoxy tryptamine hcl, 5 methoxytryptamine hydrochlorideShow MoreShow Less
PubChem CID 6198
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES COC1=CC2=C(C=C1)NC=C2CCN.Cl
Specifications
CAS Min % 96.0
CAS Max % 100.0
Melting Point 246.0°C to 248.0°C
Color Beige to White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Merck Index 14, 6002
Formula Weight 226.71
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxytryptamine hydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxytryptamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Product Content Correction
Product Title
5-Methoxytryptamine hydrochloride, 97%, Thermo Scientific™ >