Learn More
Wheaton™ 60 mL Polystyrene Sample Container
Sterile sample container offers sample security and user safety with leak free performance.
1450.00 SEK - 1770.00 SEK
Specifications
Product Type | Specimen Container |
---|---|
Closure Included | Yes |
Material | Polystyrene |
Sterility | Sterile |
Capacity (Metric) | 60 mL |
Product Code | Brand | Label | Closure Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Label | Closure Material | Price | Quantity & Availability | ||||
16414969
|
Wheaton™
360099 |
Plain | Metal |
1670.00 SEK
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16424969
|
Wheaton™
360100 |
No Label | Metal |
1770.00 SEK
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16434969
|
Wheaton™
360101 |
Printed | Metal |
1770.00 SEK
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16444969
|
Wheaton™
360102 |
No Label | Polyethylene |
1450.00 SEK
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16454969
|
Wheaton™
360103 |
Printed | Polyethylene |
1505.00 SEK
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For general sample collection, transport, and storage
- Wide container range to suit most user needs
Specifications
Specimen Container | |
Polystyrene | |
60 mL | |
Screw Cap | |
60.2 mm | |
50/Pk. |
Yes | |
Sterile | |
EU Directive 98/79/EC | |
44.2 mm | |
300/Cs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Wheaton™ 60 mL Polystyrene Sample Container