missing translation for 'onlineSavingsMsg'
Learn More

Acriflavine hydrochloride, Thermo Scientific Chemicals

An intercalating dye

696.00 SEK

Chemical Identifiers

CAS 8063-24-9
Molecular Formula C27H27Cl3N6
Molecular Weight (g/mol) 541.91
MDL Number MFCD00069039
InChI Key MKLTXAHQKDVBLY-UHFFFAOYSA-N
Synonym 3,6-Diamino-10-methylacridinium chloride hydrochloride; Euflavine
IUPAC Name 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride
SMILES Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492889
SDS View Documents
Thermo Scientific Alfa Aesar
J60048.14
25 g
696.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acriflavine Hydrochloride acts as an intercalating dye and also reported to inhibit mitochondriogenesis. It is an antiseptic and has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acriflavine Hydrochloride acts as an intercalating dye and also reported to inhibit mitochondriogenesis. It is an antiseptic and has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
A mixture of 3,6-diamino-10-methylacridinium chloride and 3,6-diaminoacridine
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acriflavine hydrochloride, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.