Learn More
Ansell ActivArmr Electrical Insulating Gloves Class 1, Black, 14 in.
ActivArmr Electrical Insulating Gloves provide the ultimate fit, comfort and performance for electrical workers' safety and protection.
Brand: Ansell RIG114B100
1121.84 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Performance Profile comfort
- Natural Rubber Latex for ultimate durability and flexibility
- Ergonomically-designed hand at rest shape for less hand fatigue
- Generous flared cuff allows room for clothing and improves ventilation
- Case hardened finish provides smoother surface for easy donning and doffing performance and protection
- Product meets all applicable NFPA, OSHA and CSA standards
- Environmentally-Friendly
- Proprietary dipping process
- ISO 14001 certified manufacturing facility
Specifications
CE Number, Category III, EN 420:2003 + A1:2009, EN 60903:2003, Class 1 | |
Specific | |
Electrical Protection Gloves | |
Applications with risk of touch voltage, Electrical Contractors, Hybrid engine manufacturing and repair, Maintenance Repairs and Operations in all industrial environments (MRO), Mass Transit Repair and Maintenance, Power maintenance, repair and transmission, Telecom repair near electrical wires | |
Beaded/Rolled | |
14 in. | |
7500/11250 |
Black | |
Natural Latex Rubber | |
10 | |
Black | |
RIG114B | |
360 mm |
Product Content Correction
Product Title
Ansell ActivArmr Electrical Insulating Gloves Class 1, Black, 14 in. > ActivArmr Electrical Insulating Gloves Class 1, Black, 14 in.; Size: 10
Spot an opportunity for improvement?Share a Content Correction