Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
Brand:  Socorex™ 825.0020
2647.00 SEK valid until 2025-07-31
Use promo code "24075" to get your promotional price.
 View more versions of this product
Product Code. 10445842
  • 3570.00 SEK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
Estimated Shipment: 13-08-2025
Description
  • Constructed of polypropylene
  • With precision setting mechanism
  • Single-handed volume setting
Specifications
Specifications
Autoclavable
825
Acura
2 to 20 μL
Polypropylene
1
Micropipette
1 Each
Certificates
View accessory products
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-07-31
    Days
    22
    Hours
    0
    Minutes
    28
    Seconds
    55
Product Content Correction
Product Title
Socorex™ Acura Variable Volume Manual Pipettes > Volume: 2 to 20μL