missing translation for 'onlineSavingsMsg'
Learn More

Hettich™ Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890

Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life

Brand:  Hettich™ 4439

2247.01 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12043914

  • 2425.00 SEK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications

Specifications

Rotor Adapter
4 x 50 mL
100 mm
4
Round Bottom
34 mm
Carriers no. 4254, 4547-B, 4579-A, 4890
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hettich™ Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890 > 4 x 50mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.