Learn More
Xylem AG Multi Refractometer Calibration Fluid 5 x 5 mL (UKAS/NIST)
Calibration fluid for calibration of refractometers.
1741.46 SEK
Specifications
Type | Calibration Fluid |
---|---|
Refractive Index | Mixed |
For Use With (Equipment) | Refractometer |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
30484474
|
Xylem
90-910 |
Soda |
1741.46 SEK
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30484454
|
Xylem
90-911 |
Beverage (USA) |
1741.46 SEK
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30484476
|
Xylem
90-912 |
Juice |
1741.46 SEK
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30484471
|
Xylem
90-913 |
Pharma and Chem |
1741.46 SEK
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30484479
|
Xylem
90-914 |
Petrochemical |
1741.46 SEK
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Calibration Fluid
- For Refractometers
Specifications
Calibration Fluid | |
Mixed | |
Refractometer |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Xylem AG Multi Refractometer Calibration Fluid 5 x 5 mL (UKAS/NIST)