Learn More
AGRP Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21249-1-AP-150UL
Description
The AGRP antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human AGRP. This antibody recognizes human antigen. The AGRP antibody has been validated for the following applications: IHC,ELISA analysis.
Specifications
AGRP | |
Polyclonal | |
Unconjugated | |
150 μL | |
181 | |
Liquid |
Immunohistochemistry, ELISA | |
350 μg/mL | |
Rabbit | |
Primary | |
Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.