missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGRP Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | AGRP |
---|---|
Concentration | 350 ÎĽg/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890865
|
Proteintech
21249-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880865
|
Proteintech
21249-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The AGRP antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human AGRP. This antibody recognizes human antigen. The AGRP antibody has been validated for the following applications: IHC,ELISA analysis.
Specifications
AGRP | |
Immunohistochemistry, ELISA | |
Unconjugated | |
Rabbit | |
181 |
350 ÎĽg/mL | |
Polyclonal | |
Liquid | |
Human | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AGRP Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech