Learn More
AGRP Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21249-1-AP-150UL
 View more versions of this product
Product Code. 16880865
  • 4652.90 SEK / 150µL
Description
The AGRP antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human AGRP. This antibody recognizes human antigen. The AGRP antibody has been validated for the following applications: IHC,ELISA analysis.
Specifications
Specifications
AGRP
Polyclonal
Unconjugated
150 μL
181
Liquid
Immunohistochemistry, ELISA
350 μg/mL
Rabbit
Primary
Human
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
AGRP Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL