Learn More
AHCYL2 Mouse anti-Human, Unconjugated, Clone: 1D1F6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67484-1-IG-20UL
 View more versions of this product
Product Code. 16880136
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The AHCYL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AHCYL2. This antibody recognizes human antigen. The AHCYL2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
AHCYL2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1D1F6
Unconjugated
20 μL
23382
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AHCYL2 Mouse anti-Human, Unconjugated, Clone: 1D1F6, Proteintech > 20 μL