Learn More
AKR1B1 Mouse anti-Human, Unconjugated, Clone: 2F11G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67498-1-IG-20UL
Description
The AKR1B1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AKR1B1. This antibody recognizes human, mouse antigen. The AKR1B1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
AKR1B1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot | |
2F11G5 | |
Unconjugated | |
20 μL | |
231 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.