Learn More
AKR1B1 Mouse anti-Human, Unconjugated, Clone: 2F11G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67498-1-IG-20UL
 View more versions of this product
Product Code. 16800166
  • 1645.54 SEK / 20µL
Description
The AKR1B1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AKR1B1. This antibody recognizes human, mouse antigen. The AKR1B1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
AKR1B1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse
Western Blot
2F11G5
Unconjugated
20 μL
231
IgG1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
AKR1B1 Mouse anti-Human, Unconjugated, Clone: 2F11G5, Proteintech > 20 μL