Learn More
AKR1B1 Mouse anti-Human, Unconjugated, Clone: 2F11G5, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | AKR1B1 |
---|---|
Clone | 2F11G5 |
Concentration | 500 ÎĽg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800166
|
Proteintech
67498-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890156
|
Proteintech
67498-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The AKR1B1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AKR1B1. This antibody recognizes human, mouse antigen. The AKR1B1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
AKR1B1 | |
500 ÎĽg/mL | |
Monoclonal | |
Mouse | |
231 | |
Primary |
2F11G5 | |
Western Blot | |
Unconjugated | |
Human, Mouse | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.