Learn More
STEMCELL Technologies ALDEFLUOR™ Assay Buffer
Additional assay buffer for use with the ALDEFLUOR™ kit
POA - 3292.65 SEK
Specifications
For Use With (Application) | Cancer Research; Neuroscience; Stem Cell Biology |
---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17157711
![]() |
STEMCELL Technologies
01701 |
25 mL | |||||||
17167711
![]() |
STEMCELL Technologies
01702 |
55 mL |
3292.65 SEK
55mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ALDEFLUOR™ Assay Buffer can be used when counterstaining ALDEFLUOR™-reacted cells with immunofluorescently-labeled antibodies
- The buffer is necessary to maintain ALDEFLUOR™ reaction product inside the cell, allowing detection of the ALDH-bright population
- While there is sufficient ALDEFLUOR™-buffer contained within the ALDEFLUOR™ kit, additional buffer may be required for certain immunolabeling procedures
Specifications
Cancer Research; Neuroscience; Stem Cell Biology |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.