Learn More
STEMCELL Technologies ALDEFLUOR™ Assay Buffer
Additional assay buffer for use with the ALDEFLUOR™ kit
Encompass_Preferred
POA - 3292.65 SEK
Specifications
For Use With (Application) Cancer Research; Neuroscience; Stem Cell Biology
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17157711
Encompass_Preferred
View Documents
STEMCELL Technologies
01701
25 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17167711
Encompass_Preferred
View Documents
STEMCELL Technologies
01702
55 mL
3292.65 SEK
55mL
View alternative products
Save Up to 
»
Description
Description
  • ALDEFLUOR™ Assay Buffer can be used when counterstaining ALDEFLUOR™-reacted cells with immunofluorescently-labeled antibodies
  • The buffer is necessary to maintain ALDEFLUOR™ reaction product inside the cell, allowing detection of the ALDH-bright population
  • While there is sufficient ALDEFLUOR™-buffer contained within the ALDEFLUOR™ kit, additional buffer may be required for certain immunolabeling procedures
Specifications
Specifications
Cancer Research; Neuroscience; Stem Cell Biology
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies ALDEFLUOR™ Assay Buffer