missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH3B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19446-1-AP-150UL
This item is not returnable.
View return policy
Description
The aldehyde dehydrogenases are a family of isozymes that may play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular gene spans about 20 kb of genomic DNA and is composed of 9 coding exons. The gene is highly expressed in kidney and lung. The functional significance of this gene as well as the cellular localization of its product are presently unknown. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
ALDH3B1 | |
Polyclonal | |
Unconjugated | |
ALDH3B1 | |
Aldehyde dehydrogenase 7, ALDH3B1, ALDH4, ALDH7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221, 309147, 67689 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43353, Q5XI42, Q80VQ0 | |
Aldh3b1 | |
ALDH3B1 Fusion Protein Ag7025 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ALDH3B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction