missing translation for 'onlineSavingsMsg'
Learn More

Hach Pacific Pty Ltd Alkaline Cyanide Reagent

For determination of low level manganese by the PAN method.

Brand:  Hach Pacific Pty Ltd 2122326

Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg

 View more versions of this product

Product Code. 16885663

  • 466.00 SEK / 50mL

Estimated Shipment: 14-04-2023
to see stock.
Add to basket

Description

Description

For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.

    Specifications

  • Method Name: PAN
  • Platform: Solution
Specifications

Specifications

50 mL
Solution
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers