Learn More
Hach Pacific Pty Ltd Alkaline Cyanide Reagent
For determination of low level manganese by the PAN method.
656.00 SEK
Specifications
Type Solution
View More Specs
Products 1
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
16885663
View Documents
Hach Pacific Pty Ltd
2122326
50 mL
656.00 SEK
50mL
Description
Description
For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.
    Specifications
  • Method Name: PAN
  • Platform: Solution
Specifications
Specifications
Solution
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hach Pacific Pty Ltd Alkaline Cyanide Reagent