Learn More
Hach Pacific Pty Ltd Alkaline Cyanide Reagent
For determination of low level manganese by the PAN method.
656.00 SEK
Description
For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.
- Specifications
- Method Name: PAN
- Platform: Solution
Specifications
Solution |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hach Pacific Pty Ltd Alkaline Cyanide Reagent