missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha 1 microglobulin Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26716-1-AP-150UL
This item is not returnable.
View return policy
Description
The Alpha 1 microglobulin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Alpha 1 microglobulin. This antibody recognizes human antigen. The Alpha 1 microglobulin antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
Alpha 1 microglobulin | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Antigen affinity purification | |
RUO | |
259 | |
Store at -20°C. Stable for one year after shipment. | |
Polyclonal Antibody | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
1000 ÎĽg/mL | |
EDC1; HCP; HI30; IATIL; ITI; ITIL; ITILC; UTI | |
Recombinant Protein | |
150 ÎĽL | |
Primary | |
Human | |
30 kDa | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Alpha 1 microglobulin Rabbit Polyclonal Antibody, Proteintech > Unconjugated; 150 ÎĽL
Spot an opportunity for improvement?Share a Content Correction