missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha 1 microglobulin Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
1567.35 SEK - 4240.22 SEK
Specifications
Antigen | Alpha 1 microglobulin |
---|---|
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30235095
|
Proteintech
26716-1-AP-20UL |
20 ÎĽL |
1567.35 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30235106
|
Proteintech
26716-1-AP-150UL |
150 ÎĽL |
4240.22 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Alpha 1 microglobulin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Alpha 1 microglobulin. This antibody recognizes human antigen. The Alpha 1 microglobulin antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
Alpha 1 microglobulin | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
EDC1; HCP; HI30; IATIL; ITI; ITIL; ITILC; UTI | |
IgG | |
Antigen affinity purification | |
Polyclonal Antibody |
1000 ÎĽg/mL | |
Polyclonal | |
Liquid | |
RUO | |
259 | |
Recombinant Protein | |
Primary | |
Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Alpha 1 microglobulin Rabbit Polyclonal Antibody, Proteintech