Learn More
Alpha SNAP Mouse anti-Human, Unconjugated, Clone: 1E2A2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67323-1-IG-150UL
 View more versions of this product
Product Code. 16879885
  • 4652.90 SEK / 150µL
Description
The Alpha SNAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Alpha SNAP. This antibody recognizes human, pig, mouse, rat antigen. The Alpha SNAP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Alpha SNAP
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1E2A2
Unconjugated
150 ÎĽL
8775
IgG2b
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Alpha SNAP Mouse anti-Human, Unconjugated, Clone: 1E2A2, Proteintech > 150 ÎĽL