Learn More
Alpha SNAP Mouse anti-Human, Unconjugated, Clone: 1E2A2, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Alpha SNAP |
---|---|
Clone | 1E2A2 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889885
|
Proteintech
67323-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879885
|
Proteintech
67323-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Alpha SNAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Alpha SNAP. This antibody recognizes human, pig, mouse, rat antigen. The Alpha SNAP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Alpha SNAP | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
8775 | |
Primary |
1E2A2 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.