Learn More
Ansell AlphaTec™ 1800 Ts PLUS
Lightweight coveralls with breathable microporous laminate technology and Type 4/5/6 protection.
Brand:  Ansell WH18T-00156-02
5399.80 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16884740
  • 7995.00 SEK / Pack of 35
Only null left
Estimated Shipment: 06-11-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • 3 piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
  • Finger Loops
  • Chinstrap
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Small
White
Hood and Socks
53 gsm
Microporous Non-woven Polyethylene Laminate
Stitched, Taped
Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders
W18T156
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 1800 Ts PLUS > Size: Small, Style: W18T156