Learn More
Ansell AlphaTec™ 4000 APOLLO
Fully encapsulated chemical protective and liquid tight suit with Type 3/4 protection.
11739.20 SEK
Specifications
Color | Green |
---|---|
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
Material | Multi-Layer Non-Woven Barrier Laminate Fabric |
Seam Type | Welded and Taped |
Style | G40T126B |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17120665
|
Ansell
GR40T-00126-07 |
3X-Large |
11739.20 SEK
Pack of 4 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
17110665
|
Ansell
GR40T-00126-02 |
Small |
11739.20 SEK
Pack of 4 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
Description
- Designed to protect, rear entry double zip system, attached socks with boot overflap
- Rear mounted breathing apparatus (BA) pouch - universal fit with most BA
- Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
- Attached Ansell Barrier™ gloves, exhalation valves and clear face visor
Specifications
Green | |
Multi-Layer Non-Woven Barrier Laminate Fabric | |
G40T126B |
Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes | |
Welded and Taped | |
103 gsm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell AlphaTec™ 4000 APOLLO