Learn More
BRAND™ Amber USP Measuring Flask
With polypropylene stopper
Brand: BRAND™ 957489
832.98 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Polypropylene | |
250 mL | |
Ground Joint NS 14/23 |
Amber | |
Borosilicate Glass (3.3) | |
A |
Product Content Correction
Product Title
BRAND™ Amber USP Measuring Flask > Capacity (Metric): 250 mL
Spot an opportunity for improvement?Share a Content Correction