Learn More
AMIGO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21874-1-AP-150UL
 View more versions of this product
Product Code. 16881425
  • 4652.90 SEK / 150µL
Description
Promotes growth and fasciculation of neurites from cultured hippocampal neurons. May be involved in fasciculation as well as myelination of developing neural axons. May have a role in regeneration as well as neural plasticity in the adult nervous system. May mediate homophilic as well as heterophilic cell-cell interaction and contribute to signal transduction through its intracellular domain.
Specifications
Specifications
AMIGO1
Polyclonal
Unconjugated
AMIGO1
ALI2, Alivin 2, AMIGO, AMIGO 1, AMIGO1, Amphoterin induced protein 1, KIAA1163
Rabbit
Antigen Affinity Chromatography
RUO
229715, 295365, 57463
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80ZD7, Q80ZD8, Q86WK6
Amigo1
AMIGO1 Fusion Protein Ag16226
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
AMIGO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated