missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMMECR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24687-1-AP-20UL
This item is not returnable.
View return policy
Description
The exact function of this gene is not known, however, submicroscopic deletion of the X chromosome including this gene, COL4A5, and FACL4 genes, result in a contiguous gene deletion syndrome, the AMME complex (Alport syndrome, mental retardation, midface hypoplasia, and elliptocytosis). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
AMMECR1 | |
Polyclonal | |
Unconjugated | |
AMMECR1 | |
AMMECR1, AMMERC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56068, 9949 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JHT5, Q9Y4X0 | |
Ammecr1 | |
AMMECR1 Fusion Protein Ag16977 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AMMECR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction