missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin VI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12542-1-AP-20UL
This item is not returnable.
View return policy
Description
Annexin VI belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. The annexin VI gene is approximately 60 kbp long and contains 26 exons. It encodes a protein of about 68 kDa that consists of eight 68-amino acid repeats separated by linking sequences of variable lengths. It is highly similar to human annexins I and II sequences, each of which contain four such repeats. Annexin VI has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis. Alternatively spliced transcript variants have been described.Specifications
Annexin VI | |
Polyclonal | |
Unconjugated | |
ANXA6 | |
67 kDa calelectrin, Annexin 6, annexin A6, Annexin VI, ANX6, ANXA6, Calphobindin II, CBP68, Chromobindin 20, CPB II, Lipocortin VI, p68, p70, Protein III | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11749, 309, 79125 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08133, P14824, P48037 | |
Anxa6 | |
Annexin VI Fusion Protein Ag3263 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Annexin VI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction