Learn More
ANO2 Mouse anti-Human, Unconjugated, Clone: 2F12G1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67638-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16890376
  • 3665.00 SEK / 150µL
Estimated Shipment: 30-03-2023
to see stock.
Description
Description
The ANO2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human ANO2. This antibody recognizes human, mouse antigen. The ANO2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ANO2
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Human
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
2F12G1
Unconjugated
150 μL
57101
IgG2a