missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANO2 Mouse anti-Human, Unconjugated, Clone: 2F12G1, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ANO2 |
---|---|
Clone | 2F12G1 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800386
|
Proteintech
67638-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890376
|
Proteintech
67638-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ANO2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human ANO2. This antibody recognizes human, mouse antigen. The ANO2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ANO2 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
57101 | |
Primary |
2F12G1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ANO2 Mouse anti-Human, Unconjugated, Clone: 2F12G1, Proteintech